![Carbamazepine-(carboxamide-13C,15N) 98 atom % 15N, 99 atom % 13C](/deepweb/assets/sigmaaldrich/product/structures/337/624/a6b83311-e1fe-4b31-93cc-3e8aeb3623cc/640/a6b83311-e1fe-4b31-93cc-3e8aeb3623cc.png)
Carbamazepine-(carboxamide-13C,15N)
![Carbamazepine-(carboxamide-13C,15N) 98 atom % 15N, 99 atom % 13C](/deepweb/assets/sigmaaldrich/product/structures/337/624/a6b83311-e1fe-4b31-93cc-3e8aeb3623cc/640/a6b83311-e1fe-4b31-93cc-3e8aeb3623cc.png)
Empirical Formula (Hill Notation):
13CC14H1215NNO
CAS Number:
Molecular Weight:
238.25
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing