Skip to Content
Merck
Calcitonin Gene Related Peptide Fragment 8-37 human ≥97% (HPLC), powder

Calcitonin Gene Related Peptide Fragment 8-37 human

Calcitonin Gene Related Peptide Fragment 8-37 human ≥97% (HPLC), powder
Synonym(s):
α-CGRP 8-37, FBHuman
Empirical Formula (Hill Notation):
C139H230N44O38
CAS Number:
Molecular Weight:
3125.59

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing