Skip to Content
Merck
BH3I-1 ≥97% (HPLC), powder, yellow

BH3I-1

BH3I-1 ≥97% (HPLC), powder, yellow
Synonym(s):
5-(p-Bromobenzylidine)-α-isopropyl-4-oxo-2-thioxo-3-thiozolidineacetic acid
Empirical Formula (Hill Notation):
C15H14BrNS2O3
CAS Number:
Molecular Weight:
400.31

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing