Skip to Content
Merck
vibrant-m

Atenolol Related Compound B

vibrant-m
Synonym(s):
2-[4-(2,3-Dihydroxypropoxy)phenyl]acetamide
Empirical Formula (Hill Notation):
C11H15NO4
CAS Number:
Molecular Weight:
225.24

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Showing 1-1 of 1