Skip to Content
Merck
Anti-γ-Tubulin antibody produced in rabbit IgG fraction of antiserum, buffered aqueous solution

Anti-γ-Tubulin antibody produced in rabbit

Anti-γ-Tubulin antibody produced in rabbit IgG fraction of antiserum, buffered aqueous solution
Synonym(s):
Anti-γ-Tubulin

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique