![Anti -Dinitrophenyl antibody, Mouse monoclonal IgE isotype, ~1 mg/mL, clone SPE-7, affinity purified immunoglobulin, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/393/692/68d4eb72-0546-483f-923d-bf2368ce24d3/640/68d4eb72-0546-483f-923d-bf2368ce24d3.jpg)
Anti -Dinitrophenyl antibody, Mouse monoclonal
![Anti -Dinitrophenyl antibody, Mouse monoclonal IgE isotype, ~1 mg/mL, clone SPE-7, affinity purified immunoglobulin, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/393/692/68d4eb72-0546-483f-923d-bf2368ce24d3/640/68d4eb72-0546-483f-923d-bf2368ce24d3.jpg)
Synonym(s):
Anti-Dinitrophenyl, Monoclonal Anti-DNP
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing