Skip to Content
Merck
All Photos(1)

Key Documents

TA9H93ED72F0

Targetmol Chemicals Inc.

Neoglycyrol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
366.37
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

3,9-dihydroxy-1-methoxy-2-(3-methyl-2-butenyl)-6H-[1]benzofuro[3,2-c]chromen-6-one

Inchi Code

1S/C21H18O6/c1-10(2)4-6-12-14(23)9-16-18(19(12)25-3)20-17(21(24)27-16)13-7-5-11(22)8-15(13)26-20/h4-5,7-9,22-23H,6H2,1-3H3

InChI key

LWESBHWAOZORCQ-UHFFFAOYSA-N

storage temp.

-80/-20

Purity

98.39%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/Neoglycyrol

MSDS

https://www.targetmol.com/compound/Neoglycyrol


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service