Skip to Content
Merck
All Photos(1)

Key Documents

C238

Sigma-Aldrich

S(−)-Cyanopindolol hemifumarate salt

solid, ≥98% (HPLC)

Synonym(s):

4-[3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-1H-indole-2-carbonitrile hemifumarate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$279.00

$279.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$279.00

About This Item

Empirical Formula (Hill Notation):
C16H21N3O2 · 0.5 C4H4O4
CAS Number:
Molecular Weight:
345.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$279.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

optical activity

[α]20/D −13.0°, c = 0.215 in methanol(lit.)

color

white to off-white

solubility

DMSO: >10 mg/mL

SMILES string

OC(=O)\C=C\C(O)=O.CC(C)(C)NC[C@H](O)COc1cccc2[nH]c(cc12)C#N.CC(C)(C)NC[C@H](O)COc3cccc4[nH]c(cc34)C#N

InChI

1S/2C16H21N3O2.C4H4O4/c2*1-16(2,3)18-9-12(20)10-21-15-6-4-5-14-13(15)7-11(8-17)19-14;5-3(6)1-2-4(7)8/h2*4-7,12,18-20H,9-10H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;;2-1+/t2*12-;/m00./s1

InChI key

ZSBITJKBOWVCCI-SODPCTSLSA-N

Biochem/physiol Actions

5-HT1A/1B serotonin receptor antagonist; β3-adrenoceptor antagonist.

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Pauwels et al.
The Journal of pharmacology and experimental therapeutics, 270(3), 938-945 (1994-09-01)
Functional activity of 5-hydroxytryptamine (5-HT) receptor ligands and beta adrenergic receptor antagonists was studied at 5-HT1B receptor sites in Chinese hamster lung (CHL) fibroblasts by measuring two cellular responses: inhibition of forskolin-stimulated cyclic AMP formation and potentiation of basic fibroblast
H Giles et al.
British journal of pharmacology, 117(6), 1119-1126 (1996-03-01)
1. Chinese hamster ovary (CHO) cells have been reported to be devoid of 5-HT receptors and have frequently been used as hosts for the expression of cloned 5-HT receptors. Unexpectedly, 5-HT was found to induce profound inhibition of forskolin-stimulated cyclic
A J Hoey et al.
British journal of pharmacology, 119(3), 564-568 (1996-10-01)
1. Cyanopindolol (CYP) is a potent antagonist at the beta 3-adrenoceptor in rat ileum. Several analogues of CYP and pindolol were synthesized that also produced antagonist effects at the beta 3-adrenoceptor. However, at high concentrations, these compounds appear to act
Yousef Risha et al.
Biomedicines, 9(2) (2021-01-28)
Membrane-derived extracellular vesicles, referred to as microvesicles (MVs), have been proposed to participate in several cancer diseases. In this study, MV fractions were isolated by differential ultracentrifugation from a metastatic breast cancer (BC) cell line MDA-MB-231 and a non-cancerous breast

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service