Skip to Content
Merck
All Photos(1)

Key Documents

KEY333430395

Key Organics / BIONET

methyl 3-amino-3-(4-chlorophenyl)propanoate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
250.12
MDL number:
Pricing and availability is not currently available. Contact Technical Service

mp

127 - 129

Iupac Name

methyl 3-amino-3-(4-chlorophenyl)propanoate hydrochloride

Inchi Code

1S/C10H12ClNO2.ClH/c1-14-10(13)6-9(12)7-2-4-8(11)5-3-7;/h2-5,9H,6,12H2,1H3;1H

InChI key

CKHLTQXMMBPPQU-UHFFFAOYSA-N

storage temp.

Store under inert gas, Hygroscopic

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/HF-0207

Melting Point

127 - 129

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/HF-0207


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service