Milli-Q® HR 7000 Connected, High-Throughput Central Water Purification Systems
High-flow Reverse Osmosis (RO) Water
The Milli-Q® HR 7000 series of connected, high-throughput systems are designed to deliver a powerful, versatile and modern central pure water solution. From a single laboratory to an entire research facility, the Milli-Q® HR 7000 range has the capacity to purify up to 13,000 L/day from tap water, reliably feeding all your type 3 water needs. This includes taps on benches, instruments, point-of-use polishers, and ultrapure water purification systems.
Features & benefits
- Advanced purification technologies for constant flow rate and reduced water consumption
- Superior ergonomic design for quick and easy maintenance
- An interactive touchscreen with helpful wizards for ease of operation and maintenance
- Modern data management capabilities for greater accessibility and data traceability
- MyMilli-Q™ Remote Care online monitoring and service capability for maximum lab productivity
Improved Sustainability in RO Water Purification
With advanced and complementary purification technologies, the Milli-Q® HR 7000 series yields reliably pure water at a constant flow rate and with reduced water consumption.
Tap water is first pretreated according to your facility’s source water quality. Progard® cartridges efficiently remove particles, colloids, free chlorine and hardness. Additional carbon and ultrafiltration systems may be supplemented as per your feed water quality.
Advanced reverse osmosis (RO) then removes 95-99% of ions, and 99% of all dissolved organics (>200 Da), microorganisms and particles.
Milli-Q® HR 7000 systems are equipped with our patented E.R.A.® (Evolutive Reject Adjustment) technology to save you water, time and money. E.R.A.® automatically assesses feed water quality to produce a constant flow rate and optimize water recovery (between 45 and 75%). The result is reduced water consumption—by up to 50% compared to other RO systems of similar throughput—and less time needed to maintain the system as there is no need for manual valve adjustments due to temperature fluctuations. E.R.A.® also prolongs the lifetime of the RO cartridge for reduced consumables waste.
Versatile configuration
Depending on daily volume needs, Milli-Q® HR 7000 systems are available in production flow rates of 60, 120, 170 or 220 L/h. Achieve higher throughput by linking up to 3 units to reach up to 13,000 L/day of Type 3 water from potable tap water.
Milli-Q® HR 7000 systems can be adapted and integrated into almost any lab or building configuration. The systems are capable of driving and controlling all ancillary equipment needed for a complete installation, including distribution pumps and loops, alarms, monitors, UV lamps and sanitation modules.
For a compact and clean solution, the Milli-Q® HR 7000 offers plug ‘n’ play compatibility with our SDS 500 storage, protection and distribution system. This perfect pairing yields high storage and distribution flow possibilities.
There’s no need to open a user manual to maintain the system – wizards guide you through basic maintenance.
Supported by MyMilli-Q™ Remote Care Online Monitoring and Service Capability
Embedded in Milli-Q® HR 7000 systems, MyMilli-Q™ Remote Care is an online service capability that operates as an extension of Milli-Q® Services. MyMilli-Q™ Remote Care lets you securely manage your system from anywhere at any time, supporting your lab’s optimal productivity.
Key features of MyMilli-Q™ Remote Care
- 24/7 secure online access to real-time system performance data as well as quality reports archived over the last 2 years
- Rapid, online diagnostics—and even remote repair—by our service team
- Receive immediate and customized notifications of system alerts and alarms by email and SMS
- Full data traceability as well as effortless data access and retrieval capabilities facilitate accreditation and reaccreditation processes
Facilitated Data Management, Traceability & Compliance
The Milli-Q® HR 7000 system makes it easier to manage and keep records of your water data, allowing you to be more audit-ready. The system collects all running parameters, including water quality, alarms and events, water usage, and consumables. Up to 2 years of events are stored in the system, which are accessible via the lab’s network or a USB key. Generate automatic e-records and save time, money and paper. This system also facilitates the lab accreditation and re-accreditation processes (ISO 15189).
Safety
- Milli-Q® HR 7000 systems are compliant to European directives related to safety and electromagnetic compatibility. Tests according to the IEC system scheme for mutual recognition have been performed by an independent and accredited Certified Body. CB test certificates issued by the NCB can be verified on the IECEE web site
- Milli-Q® HR 7000 systems are also built using components and practices recommended by UL. Systems have been tested according to national deviation and are cUL marked. The registration can be verified on the UL web site
Download the Milli-Q® HR 7000 System Brochure
Flyer: High-flow Milli-Q® Water Systems for Industry