LC/MS Analysis of β-Lactoglobulin Tryptic Digest on a Discovery® BIO Wide Pore C18 Capillary Column
CONDITIONS
column
Discovery BIO Wide Pore C18, 15 cm × 0.5 mm I.D., 5 μm particles (65519-U)
mobile phase
[A] 0.1% TFA in water; [B] 0.1% TFA in acetonitrile
gradient
5-40% B in 70 min
flow rate
14 μL/min
column temp.
30 °C
detector
+ESI mode
injection
500 pmol (5μL) β-Lactoglobulin tryptic digest in 50 mM ammonium bicarbonate
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany