Merck
  • Zc3h12c inhibits vascular inflammation by repressing NF-κB activation and pro-inflammatory gene expression in endothelial cells.
Zc3h12c inhibits vascular inflammation by repressing NF-κB activation and pro-inflammatory gene expression in endothelial cells.
The Biochemical journal (2013-01-31)
Ling Liu, Zhou Zhou, Shengping Huang, Yanhong Guo, Yanbo Fan, Ji Zhang, Jifeng Zhang, Mingui Fu, Y Eugene Chen
ABSTRACT
Endothelial activation characterized by the expression of multiple chemokines and adhesive molecules is a critical initial step of vascular inflammation, which results in recruitment of leucocytes into the sub-endothelial layer of the vascular wall and triggers vascular inflammatory diseases such as atherosclerosis. Although inhibiting endothelial inflammation has already been well recognized as a therapeutic strategy in vascular inflammatory diseases, the therapeutic targets are still elusive. In the present study we found that Zc3h12c (zinc finger CCCH-type-containing 12C), a recently discovered CCCH zinc finger-containing protein, significantly inhibited the endothelial cell inflammatory response in vitro. Overexpression of Zc3h12c significantly attenuated TNFα (tumour necrosis factor α)-induced expression of chemokines and adhesive molecules, and thus reduced monocyte adherence to HUVECs (human umbilical vein endothelial cells). Conversely, siRNA (small interfering RNA)-mediated knockdown of Zc3h12c increased the TNFα-induced expression of chemokines and adhesive molecules in HUVECs. Furthermore, forced expression of Zc3h12c decreased TNFα-induced IKKα/β [IκB (inhibitor of nuclear factor κB) kinase α/β], IκBα phosphorylation and p65 nuclear translocation, suggesting that Zc3h12c exerted its anti-inflammatory function probably by suppressing the NF-κB (nuclear factor κB) pathway. Thus Zc3h12c is an endogenous inhibitor of TNFα-induced inflammatory signalling in HUVECs and might be a therapeutic target in vascular inflammatory diseases.