Merck
  • Insufficient microwave ablation-induced promotion of distant metastasis is suppressed by β-catenin pathway inhibition in breast cancer.
Insufficient microwave ablation-induced promotion of distant metastasis is suppressed by β-catenin pathway inhibition in breast cancer.
Oncotarget (2018-02-01)
Peng Kong, Hong Pan, Muxin Yu, Lie Chen, Han Ge, Jin Zhu, Ge Ma, Li Li, Qiang Ding, Wenbin Zhou, Shui Wang
ABSTRACT
Microwave ablation (MWA), a thermal ablation, is an effective treatment for breast cancer. However, residual breast cancer is still detected. The biological characteristics of residual breast cancer after thermal ablation remain unknown. To mimic insufficient MWA
Sigma-Aldrich
MISSION® esiRNA, targeting human CTNNB1
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
2 000,00 kr
50 μg
Please contact Customer Service for Availability
3 540,00 kr