Merck
Pentosan polysulfate binds to STRO-1
Stem cell research & therapy (2017-12-15)
Jiehua Wu, Susan Shimmon, Sharon Paton, Christopher Daly, Tony Goldschlager, Stan Gronthos, Andrew C W Zannettino, Peter Ghosh
ABSTRACT
The pharmaceutical agent pentosan polysulfate (PPS) is known to induce proliferation and chondrogenesis of mesenchymal progenitor cells (MPCs) in vitro and in vivo. However, the mechanism(s) of action of PPS in mediating these effects remains unresolved. In the present report we address this issue by investigating the binding and uptake of PPS by MPCs and monitoring gene expression and proteoglycan biosynthesis before and after the cells had been exposed to limited concentrations of PPS and then re-established in culture in the absence of the drug (MPC priming). Immuno-selected STRO-1 The kinetic studies indicated that, in culture, PPS rapidly binds to MPC surface receptors, followed by internalisation and localization within the nucleus of the cells. Following PPS-priming of MPCs and a further 48 h of culture, both cell proliferation and proteoglycan synthesis were enhanced. Reduced expression of MPC-related cell surface antigen expression was promoted by the PPS priming, and RNA sequencing analysis revealed changes in the expression of 42 genes. This study has shown that priming of MPCs with low concentrations of PPS enhanced chondrogenesis and MPC proliferation by modifying their characteristic basal gene and protein expression. These findings offer a novel approach to re-programming mesenchymal stem cells for clinical indications which require the repair or regeneration of cartilaginous tissues such as in osteoarthritis and degenerative disc disease.
Sigma-Aldrich
3-(1-Pyridinio)-1-propanesulfonate, ≥97.0% (N)