Merck
  • The role of calcination temperature on structural and luminescence behaviour of novel apatite-based Ca
The role of calcination temperature on structural and luminescence behaviour of novel apatite-based Ca
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2017-10-06)
I Perhaita, L E Muresan, D T Silipas, G Borodi, Y Karabulut, J Garcia Guinea, M Ayvacikli, N Can
ABSTRACT
A series of novel apatite-based Ca
Sigma-Aldrich
Terbium(III) nitrate pentahydrate, 99.9% trace metals basis