Merck
  • A phase 2, open-label, multi-center study of amuvatinib in combination with platinum etoposide chemotherapy in platinum-refractory small cell lung cancer patients.
A phase 2, open-label, multi-center study of amuvatinib in combination with platinum etoposide chemotherapy in platinum-refractory small cell lung cancer patients.
Oncotarget (2017-11-09)
Lauren Averett Byers, Leora Horn, Jitendra Ghandi, Goetz Kloecker, Taofeek Owonikoko, Saiama Naheed Waqar, Maciej Krzakowski, Robert J Cardnell, Junya Fujimoto, Pietro Taverna, Mohammad Azab, David Ross Camidge
ABSTRACT
Amuvatinib (MP-470) is a multi-targeted kinase inhibitor with potent activity against c-Kit, synergistic with DNA-damaging agents. We evaluated amuvatinib in combination with platinum-etoposide (EP) chemotherapy by objective response rate, survival, and tolerability in platinum-refractory small cell lung cancer (SCLC) patients. This study used a Simon 2-stage design requiring ≥3 centrally confirmed responses in the first 21 subjects. Subjects received EP with 300 mg amuvatinib orally three times daily in cycles of 21 days. A three-day amuvatinib run-in period before EP occurred in Cycle 1. Subjects received the same EP chemotherapy regimen given prior to progression/relapse. Among 23 subjects treated, we observed four PRs (17.4%) per RECIST 1.1, only two of which were centrally confirmed (8.7%, response duration 119, 151 days). Three subjects (13%) had confirmed stable disease. c-Kit H-score was ≥100 in two subjects whose respective durations of disease control were 151 and 256 days. The addition of amuvatinib to EP chemotherapy in unselected, platinum-refractory SCLC did not meet the primary endpoint of ≥3 confirmed responses in stage 1. However, high c-Kit expression in two subjects with durable disease control suggests the potential for further study of amuvatinib in SCLC patients with high c-Kit expression.