Merck
Highly potent and selective Na
Bioorganic & medicinal chemistry letters (2017-10-17)
R Ian Storer, Andy Pike, Nigel A Swain, Aristos J Alexandrou, Bruce M Bechle, David C Blakemore, Alan D Brown, Neil A Castle, Matthew S Corbett, Neil J Flanagan, David Fengas, M Scott Johnson, Lyn H Jones, Brian E Marron, C Elizabeth Payne, David Printzenhoff, David J Rawson, Colin R Rose, Thomas Ryckmans, Jianmin Sun, Jonathan W Theile, Rubben Torella, Elaine Tseng, Joseph S Warmus
ABSTRACT
The discovery and selection of a highly potent and selective Na
Sigma-Aldrich
PF-06456384 trihydrochloride, ≥98% (HPLC)