Merck
  • Differential effects of zinc exposure on male and female oysters (Crassostrea angulata) as revealed by label-free quantitative proteomics.
Differential effects of zinc exposure on male and female oysters (Crassostrea angulata) as revealed by label-free quantitative proteomics.
Environmental toxicology and chemistry (2017-03-18)
Lianzhong Luo, Qinghong Zhang, Xue Kong, Heqing Huang, Weiwei You, Caihuan Ke
ABSTRACT
Oysters accumulate Zn as an adaptation to Zn exposure; however, it is not known whether male and female oysters respond differently to Zn exposure. Proteomic and real-time polymerase chain reaction analyses were used to investigate differential responses of male and female oysters (Crassostrea angulata) to Zn exposure. After exposure to 50 μg L
Sigma-Aldrich
2-Iodopyridine, 98%