Merck
Development of Gd
Journal of biomaterials science. Polymer edition (2017-01-31)
Zhenyu Gao, Yusuke Nakanishi, Shoko Noda, Haruka Omachi, Hisanori Shinohara, Hiroyuki Kimura, Yukio Nagasaki
ABSTRACT
As novel magnetic resonance imaging (MRI) contrast agent, gadofullerene encapsulated redox nanoparticles (Gd
Sigma-Aldrich
Diethyl iminodiacetate, 98%