Merck
  • Oncogenic ZEB2 activation drives sensitivity toward KDM1A inhibition in T-cell acute lymphoblastic leukemia.
Oncogenic ZEB2 activation drives sensitivity toward KDM1A inhibition in T-cell acute lymphoblastic leukemia.
Blood (2017-01-11)
Steven Goossens, Sofie Peirs, Wouter Van Loocke, Jueqiong Wang, Mina Takawy, Filip Matthijssens, Stefan E Sonderegger, Katharina Haigh, Thao Nguyen, Niels Vandamme, Magdaline Costa, Catherine Carmichael, Filip Van Nieuwerburgh, Dieter Deforce, Oded Kleifeld, David J Curtis, Geert Berx, Pieter Van Vlierberghe, Jody J Haigh
ABSTRACT
Elevated expression of the Zinc finger E-box binding homeobox transcription factor-2 (ZEB2) is correlated with poor prognosis and patient outcome in a variety of human cancer subtypes. Using a conditional gain-of-function mouse model, we recently demonstrated that ZEB2 is an oncogenic driver of immature T-cell acute lymphoblastic leukemia (T-ALL), a heterogenic subgroup of human leukemia characterized by a high incidence of remission failure or hematological relapse after conventional chemotherapy. Here, we identified the lysine-specific demethylase KDM1A as a novel interaction partner of ZEB2 and demonstrated that mouse and human T-ALLs with increased ZEB2 levels critically depend on KDM1A activity for survival. Therefore, targeting the ZEB2 protein complex through direct disruption of the ZEB2-KDM1A interaction or pharmacological inhibition of the KDM1A demethylase activity itself could serve as a novel therapeutic strategy for this aggressive subtype of human leukemia and possibly other ZEB2-driven malignancies.
Sigma-Aldrich
MISSION® esiRNA, targeting human ZEB2