Merck
  • Escitalopram, a selective serotonin reuptake inhibitor, inhibits voltage-dependent K
Escitalopram, a selective serotonin reuptake inhibitor, inhibits voltage-dependent K
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology (2017-07-15)
Han Sol Kim, Hongliang Li, Hye Won Kim, Sung Eun Shin, Mi Seon Seo, Jin Ryeol An, Kwon-Soo Ha, Eun-Taek Han, Seok-Ho Hong, Il-Whan Choi, Grace Choi, Dae-Sung Lee, Won Sun Park
ABSTRACT
We investigated the inhibitory effect of escitalopram, a selective serotonin reuptake inhibitor (SSRI), on voltage-dependent K