Merck
  • Enzyme-free chemiluminescence immunoassay for the determination of thyroid stimulating hormone.
Enzyme-free chemiluminescence immunoassay for the determination of thyroid stimulating hormone.
Talanta (2017-05-30)
Chaeyoon Shim, Richard Chong, Ji Hoon Lee
ABSTRACT
Based on the chemiluminescent resonance energy transfer (CRET) from high-energy intermediate formed in 1,1'-oxalyldiimidazole chemiluminescence (ODI-CL) reaction to fluorescent microsphere-conjugated detection antibody, a cost-effective and highly sensitive enzyme-free sandwich immunoassay with chemiluminescence detection was developed for the early diagnosis of thyroid cancer. Thyroid stimulating hormone (TSH) primary antibody immobilized on paramagnetic bead captures rapidly TSH in human serum within 15min. Also, TSH detection antibody-conjugated yellow-green microsphere, capable of emitting green light, rapidly interacted with TSH-bound TSH primary antibody immobilized on paramagnetic bead during the 30-min incubation at room temperature. The sandwich complexes immobilized on paramagnetic in PBS buffer (pH 7.4) emit bright green light with the addition of ODI-CL reagents (e.g., H
Sigma-Aldrich
Bis(2,4,6-trichlorophenyl) oxalate, BioReagent, suitable for chemiluminescence, ≥99.0% (AT)