MilliporeSigma
  • Tagging alcohols with cyclic carbonate: a versatile equivalent of (meth)acrylate for ring-opening polymerization.
Tagging alcohols with cyclic carbonate: a versatile equivalent of (meth)acrylate for ring-opening polymerization.
Chemical communications (Cambridge, England) (2008-04-11)
Russell C Pratt, Fredrik Nederberg, Robert M Waymouth, James L Hedrick
ABSTRACT
Cyclic carbonate monomers based on a single biocompatible scaffold allow for incorporation of a wide range of functional groups into macromolecules via ring-opening polymerization.