Merck
  • Cancer-associated isocitrate dehydrogenase mutations induce mitochondrial DNA instability.
Cancer-associated isocitrate dehydrogenase mutations induce mitochondrial DNA instability.
Human molecular genetics (2016-07-19)
Joanne M Kingsbury, Nachiketha Shamaprasad, R Blake Billmyre, Joseph Heitman, Maria E Cardenas
ABSTRACT
A major advance in understanding the progression and prognostic outcome of certain cancers, such as low-grade gliomas, acute myeloid leukaemia, and chondrosarcomas, has been the identification of early-occurring mutations in the NADP
Sigma-Aldrich
L-Canavanine, ≥98% (TLC), powder, from Canavalia ensiformis