Merck
  • Time-resolved dissection of early phosphoproteome and ensuing proteome changes in response to TGF-β.
Time-resolved dissection of early phosphoproteome and ensuing proteome changes in response to TGF-β.
Science signaling (2014-07-25)
Rochelle C J D'Souza, Anna M Knittle, Nagarjuna Nagaraj, Maarten van Dinther, Chunaram Choudhary, Peter ten Dijke, Matthias Mann, Kirti Sharma
ABSTRACT
Transforming growth factor-β (TGF-β) signaling promotes cell motility by inducing epithelial-to-mesenchymal transitions (EMTs) in normal physiology and development, as well as in pathological conditions, such as cancer. We performed a time-resolved analysis of the proteomic and phosphoproteomic changes of cultured human keratinocytes undergoing EMT and cell cycle arrest in response to stimulation with TGF-β. We quantified significant changes in 2079 proteins and 2892 phosphorylation sites regulated by TGF-β. We identified several proteins known to be involved in TGF-β-induced cellular processes, such as the cytostatic response, extracellular matrix remodeling, and epithelial dedifferentiation. In addition, we identified proteins involved in other cellular functions, such as vesicle trafficking, that were not previously associated with TGF-β signaling. Although many TGF-β responses are mediated by phosphorylation of the transcriptional regulators of the SMAD family by the TGF-β receptor complex, we observed rapid kinetics of changes in protein phosphorylation, indicating that many responses were mediated through SMAD-independent TGF-β signaling. Combined analysis of changes in protein abundance and phosphorylation and knowledge of protein interactions and transcriptional regulation provided a comprehensive representation of the dynamic signaling events underlying TGF-β-induced changes in cell behavior. Our data suggest that in epithelial cells stimulated with TGF-β, early signaling is a mixture of both pro- and antiproliferative signals, whereas later signaling primarily inhibits proliferation.
Sigma-Aldrich
Anti-CDK17 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-OCIAD2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution, ab2