Merck
Knockdown of ACAT-1 reduces amyloidogenic processing of APP.
FEBS letters (2007-04-07)
Henri J Huttunen, Christopher Greco, Dora M Kovacs
ABSTRACT
Previous studies have shown that acyl-coenzyme A:cholesterol acyl transferase (ACAT), an enzyme that controls cellular equilibrium between free cholesterol and cholesteryl esters, modulates proteolytic processing of APP in cell-based and animal models of Alzheimer's disease. Here we report that ACAT-1 RNAi reduced cellular ACAT-1 protein by approximately 50% and cholesteryl ester levels by 22% while causing a slight increase in the free cholesterol content of ER membranes. This correlated with reduced proteolytic processing of APP and 40% decrease in Abeta secretion. These data show that even a modest decrease in ACAT activity can have robust suppressive effects on Abeta generation.