Merck
  • Pycnogenol Ameliorates Asthmatic Airway Inflammation and Inhibits the Function of Goblet Cells.
Pycnogenol Ameliorates Asthmatic Airway Inflammation and Inhibits the Function of Goblet Cells.
DNA and cell biology (2016-11-01)
Zhaoe Liu, Bo Han, Xing Chen, Qiaoling Wu, Lijun Wang, Gang Li
ABSTRACT
Pycnogenol
Sigma-Aldrich
MISSION® esiRNA, targeting human SOAT1