Merck
  • Preparation and use of enantioenriched allenylsilanes for the stereoselective synthesis of homopropargylic ethers.
Preparation and use of enantioenriched allenylsilanes for the stereoselective synthesis of homopropargylic ethers.
Organic letters (2007-06-15)
Ryan A Brawn, James S Panek
ABSTRACT
A convenient procedure for the synthesis of highly enantioenriched allenylsilanes by Johnson orthoester Claisen rearrangement of 1-silyl propargylic alcohols is described. Allenylsilanes are then used as carbon nucleophiles in three-component, Lewis acid mediated additions to in situ generated oxonium ions, resulting in enantioenriched homopropargylic ethers.
Sigma-Aldrich
Chloro(dimethyl)phenylsilane, 98%