- De novo synthesis of Tamiflu via a catalytic asymmetric ring-opening of meso-aziridines with TMSN3.
De novo synthesis of Tamiflu via a catalytic asymmetric ring-opening of meso-aziridines with TMSN3.
Journal of the American Chemical Society (2006-05-11)
Yuhei Fukuta, Tsuyoshi Mita, Nobuhisa Fukuda, Motomu Kanai, Masakatsu Shibasaki
PMID16683784
ABSTRACT
An asymmetric ring-opening reaction of meso-aziridines with TMSN3 was developed using a catalyst prepared from Y(OiPr)3 and chiral ligand 2 in a 1:2 ratio. Excellent enantioselectivity was realized from a wide range of substrates with a practical catalyst loading. The products were efficiently converted to enantiomerically enriched 1,2-diamines, which are versatile chiral building blocks for pharmaceuticals and chiral ligands. This reaction was applied to a catalytic asymmetric synthesis of Tamiflu, a very important anti-influenza drug containing a chiral 1,2-diamino functionality.