Merck
  • (2S,4E)-2-Hydroxy-4-octen-3-one, a Male-Produced Attractant Pheromone of the Cerambycid Beetle Tylonotus bimaculatus.
(2S,4E)-2-Hydroxy-4-octen-3-one, a Male-Produced Attractant Pheromone of the Cerambycid Beetle Tylonotus bimaculatus.
Journal of chemical ecology (2015-07-08)
Yunfan Zou, Jocelyn G Millar, J Scott Blackwood, Ryan Van Duzor, Lawrence M Hanks, Judith A Mongold-Diers, Joseph C H Wong, Ann M Ray
ABSTRACT
We report the identification of a novel pheromone structure from males of the cerambycid beetle Tylonotus bimaculatus Haldeman (Cerambycinae: Hesperophanini), a species native to eastern North America. Volatiles collected from adult males contained (2S,4E)-2-hydroxyoct-4-en-3-one (71%), (3R,4E)-3-hydroxyoct-4-en-2-one (15%), (E)-4-octen-2,3-dione (13%), and 2,3-octanedione (1.5%). Four independent field bioassays with synthetic compounds confirmed that adults of both sexes were attracted by the racemate of the major component, (E)-2-hydroxyoct-4-en-3-one. No other cerambycid species were attracted in significant numbers. Attraction of both sexes is consistent with the male-produced pheromones of many other species in the subfamily Cerambycinae, but T. bimaculatus is unusual in having a pheromone chemistry that is so far unique among species in that subfamily.