Merck
  • A de novo 7.9 Mb deletion in 22q13.2→qter in a boy with autistic features, epilepsy, developmental delay, atopic dermatitis and abnormal immunological findings.
A de novo 7.9 Mb deletion in 22q13.2→qter in a boy with autistic features, epilepsy, developmental delay, atopic dermatitis and abnormal immunological findings.
European journal of medical genetics (2010-06-15)
Chih-Ping Chen, Shuan-Pei Lin, Schu-Rern Chern, Fuu-Jen Tsai, Pei-Chen Wu, Chen-Chi Lee, Yu-Ting Chen, Wen-Ling Chen, Wayseen Wang
ABSTRACT
We report a 5-year-old boy with mental retardation, autistic features, epilepsy, developmental delay, atopic dermatitis and abnormal immunological findings, carrying a 7.9 Mb de novo deletion of chromosome 22q13.2→qter. This region contains the SHANK3, NCAPH2 and CYP2D6 genes which are associated with T-cell immune response. The present case provides evidence that 22q13 deletion syndrome may be associated with immune system dysfunction in addition to neuropsychiatric disorders.