- Synthesis and in vitro evaluation of a novel iodinated resiniferatoxin derivative that is an agonist at the human vanilloid VR1 receptor.
Synthesis and in vitro evaluation of a novel iodinated resiniferatoxin derivative that is an agonist at the human vanilloid VR1 receptor.
Bioorganic & medicinal chemistry letters (2002-04-06)
Mark E McDonnell, Sui Po Zhang, Adrienne E Dubin, Scott L Dax
PMID11934585
ABSTRACT
Using a 'directed' iodination procedure, novel iodo-resiniferatoxin congeners were synthesized from 4-acetoxy-3-methoxyphenylacetic acid and resiniferinol- 9,13,14-ortho-phenylacetate (ROPA). The 2-iodo-4-hydroxy-5-methoxyphenylacetic acid ester of resiniferinol 5 displayed high affinity binding (K(i)=0.71 nM) for the human vanilloid VR1 receptor and functioned as a partial agonist.