Merck
  • Influence of a chromophore analogue in the protein cage of a photoactive yellow protein.
Influence of a chromophore analogue in the protein cage of a photoactive yellow protein.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2015-07-17)
Norio Hamada, Zhe Tan, Yasuo Kanematsu, Naoya Inazumi, Ryosuke Nakamura
ABSTRACT
Time-resolved spectra of a photoactive yellow protein (PYP) containing cyano-p-coumaric acid (CHCA) were recorded. To understand the mechanism of photo-isomerization, an electron-withdrawing CN group was introduced into the PYP to alter the C[double bond, length as m-dash]C double bond character. Free CHCA chromophores in aqueous solution underwent photo-isomerization whereas PYP with a bound CHCA (PYP-CN) exhibited no photocycle at acidic or alkaline pH or in urea and other solutions. Furthermore, no photocycle was observed with PYP mutants after illumination. This phenomenon cannot be fully explained by the electron-withdrawing properties of the CN group. We conclude that the CHCA chromophore in PYP was locked in the protein cage and that the CN group interacted with the protein residues.
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)