Merck
  • Structural basis for processivity and antiviral drug toxicity in human mitochondrial DNA replicase.
Structural basis for processivity and antiviral drug toxicity in human mitochondrial DNA replicase.
The EMBO journal (2015-06-10)
Michal R Szymanski, Vladmir B Kuznetsov, Christie Shumate, Qingchao Meng, Young-Sam Lee, Gayatri Patel, Smita Patel, Y Whitney Yin
ABSTRACT
The human DNA polymerase gamma (Pol γ) is responsible for DNA replication in mitochondria. Pol γ is particularly susceptible to inhibition by dideoxynucleoside-based inhibitors designed to fight viral infection. Here, we report crystal structures of the replicating Pol γ-DNA complex bound to either substrate or zalcitabine, an inhibitor used for HIV reverse transcriptase. The structures reveal that zalcitabine binds to the Pol γ active site almost identically to the substrate dCTP, providing a structural basis for Pol γ-mediated drug toxicity. When compared to the apo form, Pol γ undergoes intra- and inter-subunit conformational changes upon formation of the ternary complex with primer/template DNA and substrate. We also find that the accessory subunit Pol γB, which lacks intrinsic enzymatic activity and does not contact the primer/template DNA directly, serves as an allosteric regulator of holoenzyme activities. The structures presented here suggest a mechanism for processivity of the holoenzyme and provide a model for understanding the deleterious effects of Pol γ mutations in human disease. Crystal structures of the mitochondrial DNA polymerase, Pol γ, in complex with substrate or antiviral inhibitor zalcitabine provide a basis for understanding Pol γ-mediated drug toxicity.
Sigma-Aldrich
DL-Cysteine, technical grade