Merck
  • Differential inhibition of various adenylyl cyclase isoforms and soluble guanylyl cyclase by 2',3'-O-(2,4,6-trinitrophenyl)-substituted nucleoside 5'-triphosphates.
Differential inhibition of various adenylyl cyclase isoforms and soluble guanylyl cyclase by 2',3'-O-(2,4,6-trinitrophenyl)-substituted nucleoside 5'-triphosphates.
The Journal of pharmacology and experimental therapeutics (2009-06-06)
Srividya Suryanarayana, Martin Göttle, Melanie Hübner, Andreas Gille, Tung-Chung Mou, Stephen R Sprang, Mark Richter, Roland Seifert
ABSTRACT
Adenylyl cyclases (ACs) catalyze the conversion of ATP into the second messenger cAMP and play a key role in signal transduction. In a recent study (Mol Pharmacol 70:878-886, 2006), we reported that 2',3'-O-(2,4,6-trinitrophenyl)-substituted nucleoside 5'-triphosphates (TNP-NTPs) are potent inhibitors (K(i) values in the 10 nM range) of the purified catalytic subunits VC1 and IIC2 of membranous AC (mAC). The crystal structure of VC1:IIC2 in complex with TNP-ATP revealed that the nucleotide binds to the catalytic site with the TNP-group projecting into a hydrophobic pocket. The aims of this study were to analyze the interaction of TNP-nucleotides with VC1:IIC2 by fluorescence spectroscopy and to analyze inhibition of mAC isoforms, soluble AC (sAC), soluble guanylyl cyclase (sGC), and G-proteins by TNP-nucleotides. Interaction of VC1:IIC2 with TNP-NDPs and TNP-NTPs resulted in large fluorescence increases that were differentially reduced by a water-soluble forskolin analog. TNP-ATP turned out to be the most potent inhibitor for ACV (K(i), 3.7 nM) and sGC (K(i), 7.3 nM). TNP-UTP was identified as the most potent inhibitor for ACI (K(i), 7.1 nM) and ACII (K(i), 24 nM). TNP-NTPs inhibited sAC and GTP hydrolysis by G(s)- and G(i)-proteins only with low potencies. Molecular modeling revealed that TNP-GTP and TNP-ATP interact very similarly, but not identically, with VC1:IIC2. Collectively, our data show that TNP-nucleotides are useful fluorescent probes to monitor conformational changes in VC1:IIC2 and that TNP-NTPs are a promising starting point to develop isoform-selective AC and sGC inhibitors. TNP-ATP is the most potent sGC inhibitor known so far.
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I