- Novel ATP6V0A4 mutation described in a Tunisian patient with distal renal tubular acidosis.
Novel ATP6V0A4 mutation described in a Tunisian patient with distal renal tubular acidosis.
Clinical nephrology (2012-08-03)
Donia El Hayek, Hassan Bouzidi, Gustavo Pérez de Nanclares, Habib Soua, Jemni Ben Chibani, Gema Ariceta, Luis Castaño, Amel Haj Khelil
PMID22854161
ABSTRACT
Few data regarding molecular diagnosis of primary distal renal tubular acidosis (DRTA) in Tunisian population are available. 25-day-old male patient from consanguineous parents of Tunisian origin diagnosed with DRTA and without hearing impairment observed later in life. ATP6V0A4 gene sequencing demonstrated a novel homozygous G deletion in exon 13 (c.1221delG, p.Met408CysfsX10), leading to a premature stop codon. A novel ATP6V0A4 gene mutation confirmed autosomal recessive DRTA with normal hearing in the patient. Molecular analysis may help to rapidly diagnose autosomal recessive DRTA in Tunisian population.