Merck
  • Differences in recurrent prolapse at 1 year after total vs supracervical hysterectomy and robotic sacrocolpopexy.
Differences in recurrent prolapse at 1 year after total vs supracervical hysterectomy and robotic sacrocolpopexy.
International urogynecology journal (2014-11-05)
Erinn M Myers, Lauren Siff, Blake Osmundsen, Elizabeth Geller, Catherine A Matthews
ABSTRACT
Optimal management of the cervix at the time of hysterectomy and sacrocolpopexy for primary uterovaginal prolapse is unknown. Our hypothesis was that recurrent prolapse at 1 year would be more likely after a supracervical robotic hysterectomy (SRH) compared with a total robotic hysterectomy (TRH) at the time of robotic sacrocolpopexy (RSCP) for uterovaginal prolapse. This was a retrospective cohort analysis of 83 women who underwent hysterectomy with RSCP over a 24-month period (40 with TRH and 43 with SRH). At 1 year post-procedure, subjects completed validated questionnaires regarding pelvic floor symptoms, sexual function, and global satisfaction, and underwent a pelvic examination to identify mesh exposure and evaluate pelvic floor support. Demographics of the two groups were similar, except for a higher mean body mass index in the TRH group (31.9 TRH vs 25.8 SRH kg/m(2), p < 0.001). The rate of recurrent prolapse ≥ stage II was higher for women who underwent SRH compared with TRH (41.9 % vs 20.0 %, p = 0.03; OR 2.8, 95 % CI, 1.07-7.7). However, when this was analyzed as recurrence ≥ hymen, there was no difference between groups (12.5 % TRH vs 18.6 % SRH, p = 0.45). Likewise, there was no difference between groups when a composite measure of success was used (30 out of 40 [75 %] TRH vs 29 out of 43 [67.4 %] SRH, p = 0.45). Women who underwent an SRH were 2.8 times more likely to have a recurrent prolapse, ≥ stage II, at 1 year, compared with those who underwent a TRH, but when composite assessment scores were used there was no difference between the groups.
Sigma-Aldrich
Thyrotropin releasing hormone, ≥98% (HPLC), powder