Merck
  • Application of gas chromatography/electron capture negative chemical ionization high-resolution mass spectrometry for analysis of DNA and protein adducts.
Application of gas chromatography/electron capture negative chemical ionization high-resolution mass spectrometry for analysis of DNA and protein adducts.
Chemical research in toxicology (1998-06-20)
A Ranasinghe, N Scheller, K Y Wu, P B Upton, J A Swenberg
ABSTRACT
The analytical potential of gas chromatography/electron capture negative chemical ionization high-resolution mass spectrometry (HRMS) for characterization and quantitation of DNA and hemoglobin adducts was demonstrated using three model compounds: N2, 3-ethenoguanine (EG), 7-(2-hydroxyethyl)guanine (7-HEG), and N-(2-hydroxyethyl)valine (HEV). At a resolving power of 10 000, the signal-to-noise (S/N) ratios obtained from quantitative selected ion monitoring (SIM) experiments using biological samples were comparable to or better than existing unit mass resolution experiments due to the reduction of chemical noise from the use of narrower mass windows. The specificity gained by HRMS was essential for quantitation of ultratrace amounts near the limit of detection since coeluting interferences of the analyte or internal standard can lead to inaccurate measurement of response factors. The limit of detection (LOD) was 100 amol (S/N = 5) using a pure standard of TTB2-EG. The LOD for complete assays using spiked samples was 500 amol (S/N = 5) for EG and 600 amol (S/N = 5) injected for 7-HEG. The standard deviation (SD) for the HRMS quantitative measurements was typically less than 10%. The SD for the complete biological assays as determined by spiking replicate samples was less than 15%. This method has adequate sensitivity and specificity to accurately measure DNA and protein adducts as low as endogenous concentrations in rodent and human tissues.