Merck
1-Bromo-4-methyl-2-nitro-benzene.
Acta crystallographica. Section E, Structure reports online (2011-11-08)
Ping Li, Hai Wang, Ximan Zhang, Hongyu Chen
ABSTRACT
In the title compound, C(7)H(6)BrNO(2), the dihedral angle between the nitro group and the phenyl ring is 14.9 (11)°.