Merck
  • Synthesis, DNA-binding activity and cytotoxicity of carbamate derivatives of Hoechst 33258 in breast cancer MCF-7 cells.
Synthesis, DNA-binding activity and cytotoxicity of carbamate derivatives of Hoechst 33258 in breast cancer MCF-7 cells.
Biological & pharmaceutical bulletin (2002-07-23)
Krzysztof Bielawski, Anna Bielawska, Sławomir Wołczyński
ABSTRACT
A series of carbamate derivatives of Hoechst 33258 was prepared as potential anticancer agents. These new compounds (1-4) were readily prepared in good yields by addition of chloroethyl, bromoethyl, chloropropyl or 4-(chloromethyl)phenyl isocyanates to Hoechst 33258. Their cytotoxic activity was evaluated on human breast cancer MCF-7. Compounds 1-4 were more cytotoxic than Hoechst 33258. In particular derivative 4, the most active of the series, is up to 3 times more potent than Hoechst 33258. The DNA-binding ability of these compounds was evaluated by an ultrafiltration method using calf thymus DNA. These data show that in broad terms the cytotoxic potency of 1-4 in cultured breast cancer MCF-7 cells increases, in accord with their increases in DNA affinity, as shown by the binding constant values.
Sigma-Aldrich
4-(Chloromethyl)phenyl isocyanate, 97%