Merck
Asymmetric synthesis of all stereoisomers of 6-methylpipecolic acids.
Amino acids (2003-04-23)
H Takahata, M Shimizu
ABSTRACT
Asymmetric synthesis of all four stereoisomers of 6-methylpipecolic acids with high enantiomeric purity via iterative AD reaction, starting from 1,6-heptadiene, has been described.