- Use of beta-methylphenylalanine (beta MeF) residues to probe the nature of the interaction of substance P with its receptor: effects of beta MeF-containing substance P analogs on rabbit iris smooth muscle contraction.
Use of beta-methylphenylalanine (beta MeF) residues to probe the nature of the interaction of substance P with its receptor: effects of beta MeF-containing substance P analogs on rabbit iris smooth muscle contraction.
The effects of substituting (2S,3S)-beta-methylphenylalanine (S-beta MeF) or (2S,3R)-beta-methylphenylalanine (R-beta MeF) for the Phe7 and/or Phe8 residues of the tachykinin substance P (SP, RPKPQQFFGLM-NH2) upon the ability of SP to stimulate contraction of the rabbit iris smooth muscle were investigated. The eight beta MeF-containing SP analogs (four monosubstituted analogs, four disubstituted analogs) 1-8 were synthesized and found to be agonsts of SP in the smooth muscle contraction assay, having EC50 values ranging from 0.15 to 10.0 nM. Three analogs are significantly more active than SP [8R-(beta MeF)SP (4), 7S,8S-(beta MeF)2SP (5), and 7R,8S-(beta MeF)2SP (6)], three analogs are approximately equipotent with SP [7S-(beta MeF)SP (1), 7R-(beta MeF)SP (2), and 7S,8R-(beta MeF)2SP (8)], and two analogs are significantly less active than SP [8S-(beta MeF)SP (3) and 7R,8R-(beta MeF)2SP (7)]. The effects of the beta MeF substitutions upon the activity of SP are not additive and cannot be explained using simple conformational models which focus only on the side chain conformations of the beta MeF residues. It is postulated that the beta MeF residues induce minor distortions in the peptide backbone with resultant consequences upon peptide-receptor binding which are not dictated soley by the side chain conformations. This idea is consistent with 1H-NMR data for the monosubstituted analogs 1-4, which imply that the beta MeF substitutions cause slight distortions in the peptide backbone and that the beta MeF side chains are assuming trans or gauche(-) conformations.