Merck
  • Draft genome sequence of the marine bacterium Streptomyces griseoaurantiacus M045, which produces novel manumycin-type antibiotics with a pABA core component.
Draft genome sequence of the marine bacterium Streptomyces griseoaurantiacus M045, which produces novel manumycin-type antibiotics with a pABA core component.
Journal of bacteriology (2011-05-10)
Fuchao Li, Peng Jiang, Huajun Zheng, Shengyue Wang, Guoping Zhao, Song Qin, Zhaopu Liu
ABSTRACT
Streptomyces griseoaurantiacus M045, isolated from marine sediment, produces manumycin and chinikomycin antibiotics. Here we present a high-quality draft genome sequence of S. griseoaurantiacus M045, the first marine Streptomyces species to be sequenced and annotated. The genome encodes several gene clusters for biosynthesis of secondary metabolites and has provided insight into genomic islands linking secondary metabolism to functional adaptation in marine S. griseoaurantiacus M045.
Sigma-Aldrich
Manumycin A from Streptomyces parvulus