- [Testing of some dibenzoazepines and dibenzocycloheptadiene derivatives with high pressure thin layer chromatography].
[Testing of some dibenzoazepines and dibenzocycloheptadiene derivatives with high pressure thin layer chromatography].
Acta poloniae pharmaceutica (1990-01-01)
H Szumiło, M Przyborowska, L Przyborowski, G Misztal
PMID12959235
ABSTRACT
An influence of several adsorbents and the composition of the eluent mixtures on the separation of dibenzoazepine and dibenzocycloheptadiene derivatives was examined by thin-layer chromatography. The best system (RP-18 and methanol-buffer solution pH 8.5 19:1) was employed for the separation of amitryptiline and doxepine by HPLC. Both compounds were determined in ng amounts in pharmaceutical preparations and in blood by addition and in patients blood. Doxepine was used as internal standard for the determination of amitryptiline.