Merck
  • Cumulative and antagonistic effects of a mixture of the antiandrogens vinclozolin and iprodione in the pubertal male rat.
Cumulative and antagonistic effects of a mixture of the antiandrogens vinclozolin and iprodione in the pubertal male rat.
Toxicological sciences : an official journal of the Society of Toxicology (2009-07-01)
Chad R Blystone, Christy S Lambright, Mary C Cardon, Johnathan Furr, Cynthia V Rider, Phillip C Hartig, Vickie S Wilson, Leon E Gray
ABSTRACT
Vinclozolin and iprodione are dicarboximide fungicides that display antiandrogenic effects in the male rat, which suggests that a mixture would lead to cumulative effects on androgen-sensitive end points. Iprodione is a steroid synthesis inhibitor, but androgen receptor antagonist activity, which is displayed by vinclozolin, has not been fully evaluated. Here, we demonstrate that iprodione binds to the human androgen receptor (IC(50) = 86.0 microM), reduces androgen-dependent gene expression, and reduces androgen-sensitive tissue weights in castrated male rats (Hershberger assay). Since vinclozolin and iprodione affect common targets in the pubertal male rat, we tested the hypothesis that a mixture would have cumulative antiandrogenic effects. An iprodione dose, that does not significantly affect androgen-dependent morphological end points, was combined with vinclozolin doses (2 x 5 factorial design). Sprague-Dawley rats were dosed by gavage with vinclozolin at 0, 10, 30, 60, and 100 mg/kg/day with and without 50 mg iprodione/kg/day from postnatal day (PND) 23 to 55-57 (n = 8 per group). The age at puberty (preputial separation [PPS]), organ weights, serum hormones, and ex vivo testis steroid hormone production were measured. Vinclozolin delayed PPS, reduced androgen-sensitive organ weights, and increased serum testosterone. The addition of iprodione enhanced the vinclozolin inhibition of PPS (PND 47.5 vs.49.1; two-way ANOVA: iprodione main effect p = 0.0002). The dose response for several reproductive and nonreproductive organ weights was affected in a cumulative manner. In contrast, iprodione antagonized the vinclozolin-induced increase in serum testosterone. These results demonstrate that these fungicides interact on common targets in a tissue-specific manner when coadministered to the pubertal male rat.
Product Description
Supelco
Iprodione, PESTANAL®, analytical standard
Sigma-Aldrich
[3-(3,5-Dichlorophenyl)-2,4-dioxoimidazolidinyl]-N-(methylethyl)carboxamide, 97%