Merck
  • Studies on esterification and sulfonation of riboflavin via semi-empirical methods.
Studies on esterification and sulfonation of riboflavin via semi-empirical methods.
Acta poloniae pharmaceutica (2013-08-08)
Wojciech Pajak, Elzbieta Brzezińska, Cecylia Mielczarek
ABSTRACT
The article presents extended computer investigations of various sulfate derivatives of riboflavin. A number of physicochemical parameters such as total energy, binding energy and formation heat were calculated via semi-empirical methods AM1 and PM3 for the different derivatives of riboflavin. Their analysis made it possible to determine the sequence of formation of sulfate derivatives--esterification is the easiest at hydroxyl groups at the farthest positions from the ring. This methodology may be used to study biologically active compounds.
Product Description
Supelco
Riboflavin (B2), analytical standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%