Merck
  • Synthesis of functionalized spiroindolines via palladium-catalyzed methine C-H arylation.
Synthesis of functionalized spiroindolines via palladium-catalyzed methine C-H arylation.
Organic letters (2013-03-05)
Tanguy Saget, David Perez, Nicolai Cramer
ABSTRACT
The synthesis of cyclopropyl spiroindolines is described using an intramolecular palladium(0)-catalyzed C-H functionalization of a methine C(sp(3))-H bond. This transformation can be coupled with intermolecular Suzuki couplings or direct arylations of heteroaromatics to access functionalized indoline scaffolds in a single step.
Sigma-Aldrich
Indoline, ReagentPlus®, 99%