Merck
  • Relationship between bradykinin-induced relaxation and endogenous epoxyeicosanoid synthesis in human bronchi.
Relationship between bradykinin-induced relaxation and endogenous epoxyeicosanoid synthesis in human bronchi.
American journal of physiology. Lung cellular and molecular physiology (2013-02-19)
Yacine Tabet, Marco Sirois, Chantal Sirois, Edmond Rizcallah, Éric Rousseau
ABSTRACT
Epoxyeicosanoids (EETs) are produced by cytochrome P-450 epoxygenase; however, it is not yet known what triggers their endogenous production in epithelial cells. The relaxing effects of bradykinin are known to be related to endogenous production of epithelial-derived hyperpolarizing factors (EpDHF). Because of their effects on membrane potential, EETs have been reported to be EpDHF candidates (Benoit C, Renaudon B, Salvail D, Rousseau E. Am J Physiol Lung Cell Mol Physiol 280: L965-L973, 2001.). Thus, we hypothesized that bradykinin (BK) may stimulate endogenous EET production in human bronchi. To test this hypothesis, the relaxing and hyperpolarizing effects of BK and 14,15-EET were quantified on human bronchi, as well as the effects of various enzymatic inhibitors on these actions. One micromolar BK or 1 μM 14,15-EET induced a 45% relaxation on the tension induced by 30 nM U-46619 [a thromboxane-prostanoid (TP)-receptor agonist]. These BK-relaxing effects were reduced by 42% upon addition of 10 nM iberiotoxin [a large-conductance Ca(2+)-sensitive K(+) (BK(Ca)) channel blocker], by 27% following addition of 3 μM 14,15-epoxyeicosa-5(Z)-enoic acid (an EET antagonist), and by 32% with 3 μM N-methanesulfonyl-6-(2-propargyloxyphenyl)hexanamide (MS-PPOH, an epoxygenase inhibitor). Hence, BK and 14,15-EET display net hyperpolarizing effects on airway smooth muscle cells that are related to the activation of BK(Ca) channels and ultimately yielding to relaxation. Data also indicate that 3 μM MS-PPOH reduced the hyperpolarizing effects of BK by 43%. Together, the present data support the current hypothesis suggesting a direct relationship between BK and the production of EET regioisomers. Because of its potent anti-inflammatory and relaxing properties, epoxyeicosanoid signaling may represent a promising target in asthma and chronic obstructive pulmonary disease.
Sigma-Aldrich
Bradykinin acetate salt, powder, ≥98% (HPLC)
Sigma-Aldrich
9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F, solution, 10 mg/mL in methyl acetate