Merck
  • Point mutations within the fatty acid synthase type II dehydratase components HadA or HadC contribute to isoxyl resistance in Mycobacterium tuberculosis.
Point mutations within the fatty acid synthase type II dehydratase components HadA or HadC contribute to isoxyl resistance in Mycobacterium tuberculosis.
Antimicrobial agents and chemotherapy (2012-11-02)
Laila Gannoun-Zaki, Laeticia Alibaud, Laurent Kremer
ABSTRACT
The mechanism by which the antitubercular drug isoxyl (ISO) inhibits mycolic acid biosynthesis has not yet been reported. We found that point mutations in either the HadA or HadC component of the type II fatty acid synthase (FAS-II) are associated with increased levels of resistance to ISO in Mycobacterium tuberculosis. Overexpression of the HadAB, HadBC, or HadABC heterocomplex also produced high-level resistance. These results show that the FAS-II dehydratases are involved in ISO resistance.
Sigma-Aldrich
N-Phenylthiourea, ≥98%