Merck
  • [Large-scale fragmentation of dna and the death of tumor cells by the action of the binary system ascorbic acid-metallocomplexes of cobalt in vitro].
[Large-scale fragmentation of dna and the death of tumor cells by the action of the binary system ascorbic acid-metallocomplexes of cobalt in vitro].
Tsitologiia (2012-07-26)
A I Medvedev, V V Leshchenko
ABSTRACT
High-molecular-weight DNA fragments are the markers of the early stage of apoptosis induced in eukaryotic cells by cytotoxins of different nature. The dynamics of the development of large-scale DNA fragmentation in K-562 leukemia cells by the action of the antitumor drug, the binary system ascorbic acid--cobalt phthalocyanine within 48 h of incubation, which correspond to two periods of the doubling of cell number in growing control cultures, have been studied. It was shown that, within the first hours of incubation, hydrogen peroxide generated by the system induces the formation of DNA fragments from 2200 to 50 kbp long. Later on the cell death accompanied by a decrease in the content of fragmented DNA is observed. Within 24 h of incubation, part of fragmented DNA remains unrepaired; after 48 h of incubation, a delay or a slowed down proliferation of K-562 cells, which differ from control cells also by a high level of death and a higher content of high-molecular-weight DNA fragments, is observed.
Sigma-Aldrich
Cobalt(II) phthalocyanine, β-form, Dye content 97 %